missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Aldehyde dehydrogenase 5 Antibody - BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-92625-0.1ml
This item is not returnable.
View return policy
Description
Aldehyde dehydrogenase 5 Polyclonal antibody specifically detects Aldehyde dehydrogenase 5 in Human samples. It is validated for Western Blot, Immunocytochemistry/ Immunofluorescence
Specifications
| Aldehyde dehydrogenase 5 | |
| Polyclonal | |
| Western Blot 1:500-1:2000, Immunocytochemistry/ Immunofluorescence 1:50-1:200 | |
| aldehyde dehydrogenase 1 family, member B1, Aldehyde dehydrogenase 5, Aldehyde dehydrogenase family 1 member B1, ALDH class 2, ALDH5aldehyde dehydrogenase X, mitochondrial, ALDHXacetaldehyde dehydrogenase 5, EC 1.2.1, EC 1.2.1.3, MGC2230, mitochondrial aldehyde dehydrogenase X | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 278-517 of human ALDH1B1 (NP_000683.3). NLKRVTLELGGKSPSIVLADADMEHAVEQCHEALFFNMGQCCCAGSRTFVEESIYNEFLERTVEKAKQRKVGNPFELDTQQGPQVDKEQFERVLGYIQLGQKEGAKLLCGGERFGERGFFIKPTVFGGVQDDMRIAKEEIFGPVQPLFKFKKIEEVVERANNTRYGLAAAVFTRDLDKAMYFTQALQAGTVWVNTYNIVTCHTPFGGFKESGNGRELGEDGLKAYTEVKTVTIKVPQKNS | |
| 0.1 mL | |
| Cancer, Endocrinology, Signal Transduction | |
| 219 | |
| Store at -20°C. Avoid freeze-thaw cycles. | |
| IgG |
| Western Blot, Immunofluorescence | |
| Unconjugated | |
| PBS with 50% glycerol, pH7.3. | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction