missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ ALDH16A1 Recombinant Rabbit Monoclonal Antibody (JE62-68)

Rabbit Recombinant Monoclonal Antibody
Brand: Invitrogen™ MA544816
This item is not returnable.
View return policy
Description
ALDH16A1 Recombinant Monoclonal Antibody for Western Blot, IHC (P)
This gene encodes a member of the aldehyde dehydrogenase superfamily. The family members act on aldehyde substrates and use nicotinamide adenine dinucleotide phosphate (NADP) as a cofactor. This gene is conserved in chimpanzee, dog, cow, mouse, rat, and zebrafish. The protein encoded by this gene interacts with maspardin, a protein that when truncated is responsible for Mast syndrome. Alternatively spliced transcript variants encoding different isoforms have been found for this gene.
Specifications
| ALDH16A1 | |
| Recombinant Monoclonal | |
| 1 mg/mL | |
| TBS with 0.05% BSA, 40% glycerol and 0.05% sodium azide; pH 7.4 | |
| Q3T1L0, Q571I9, Q8IZ83 | |
| ALDH16A1 | |
| Synthetic peptide within human ALDH16A1 aa 550-600/802. | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
| Immunohistochemistry (Paraffin), Western Blot | |
| JE62-68 | |
| Unconjugated | |
| ALDH16A1 | |
| 2410004H02Rik; AI452234; aldehyde dehydrogenase 16 family member A1; aldehyde dehydrogenase 16 family, member A1; aldehyde dehydrogenase 16A1; aldehyde dehydrogenase family 16 member A1; ALDH16A1; LOW QUALITY PROTEIN: aldehyde dehydrogenase family 16 member A1; mFLJ00209 | |
| Rabbit | |
| Protein A | |
| RUO | |
| 126133, 361571, 69748 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction