missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ALDH18A1 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody has been used in 2 publications
€ 380.00 - € 698.00
Specifications
| Antigen | ALDH18A1 |
|---|---|
| Applications | Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), KnockDown |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Host Species | Rabbit |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18469410
|
Novus Biologicals
NBP1-83325-25ul |
25 μL |
€ 380.00
25µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18266108
|
Novus Biologicals
NBP1-83325 |
0.1 mL |
€ 698.00
0.10mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
ALDH18A1 Polyclonal specifically detects ALDH18A1 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin, Knockdown Validated.Specifications
| ALDH18A1 | |
| Polyclonal | |
| Rabbit | |
| Human, Mouse, Rat | |
| aldehyde dehydrogenase 18 family, member A1, Aldehyde dehydrogenase family 18 member A1, delta-1-pyrroline-5-carboxylate synthase, EC 2.1.2.11, EC 2.3.3.13, GSASdelta1-pyrroline-5-carboxlate synthetase, MGC117316, P5CSdelta-1-pyrroline-5-carboxylate synthetase, PYCSEC 1.2.1.41, pyrroline-5-carboxylate synthetase (glutamate gamma-semialdehyde synthetase) | |
| ALDH18A1 | |
| IgG | |
| Affinity Purified | |
| Specificity of human ALDH18A1 antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins. |
| Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), KnockDown | |
| Unconjugated | |
| RUO | |
| PBS (pH 7.2) and 40% Glycerol with 0.02% Sodium Azide | |
| 5832 | |
| This antibody was developed against Recombinant Protein corresponding to amino acids:SIVEQVSVLQNQGREMMLVTSGAVAFGKQRLRHEILLSQSVRQALHSGQNQLKEMAIPVLEARACAAAGQSGLMALYEAMFTQYSICAAQILVTNLDFHDEQKRRNLNGTLHELLRMNIVPIVNTNDAVVPPAEPNSDLQGVNVISVKD | |
| Primary | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title