missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ALDH1A3 Antibody, DyLight 755, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-15339IR
This item is not returnable.
View return policy
Description
ALDH1A3 Polyclonal specifically detects ALDH1A3 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen, Immunoblotting.
Specifications
| ALDH1A3 | |
| Polyclonal | |
| 50mM Sodium Borate | |
| Rabbit | |
| Immunogen affinity purified | |
| RUO | |
| Primary | |
| Human, Mouse, Rat | |
| Purified |
| Western Blot, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Immunohistochemistry (Frozen), Immunoblot | |
| DyLight 755 | |
| aldehyde dehydrogenase 1 family, member A3, Aldehyde dehydrogenase 6, aldehyde dehydrogenase family 1 member A3, ALDH1A6, ALDH6acetaldehyde dehydrogenase 6, EC 1.2.1, EC 1.2.1.5, RalDH3, RALDH-3, Retinaldehyde dehydrogenase 3 | |
| Recombinant protein encompassing a sequence within the center region of human ALDH1A3. The exact sequence is proprietary. | |
| 0.1 mL | |
| Vision | |
| 220 | |
| Store at 4°C in the dark. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction