missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ALDH1A3 Mouse anti-Canine, Human, Clone: OTI4B6, liquid, TrueMAB™
Mouse Monoclonal Antibody
Brand: Origene Technologies TA502805
This item is not returnable.
View return policy
Description
Aldehyde dehydrogenase isozymes are thought to play a major role in the detoxification of aldehydes generated by alcohol metabolism and lipid peroxidation. The enzyme encoded by this gene uses retinal as a substrate, either in a free or cellular retinol-binding protein form.Specifications
| ALDH1A3 | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with 1% BSA, 50% glycerol and 0.02% sodium azide | |
| P47895 | |
| ALDH1A3 | |
| Human recombit protein fragment corresponding to amino acids 1-100 and 413-512 of human ALDH1A3 produced in E.coli. | |
| 100 μL | |
| Primary | |
| Canine, Human | |
| Antibody | |
| IgG1 |
| Flow Cytometry, Immunohistochemistry (Paraffin), Western Blot | |
| OTI4B6 | |
| Unconjugated | |
| ALDH1A3 | |
| ALDH1A6, ALDH6, MCOP8, RALDH3 | |
| Mouse | |
| Affinity Chromatography | |
| RUO | |
| 100856346, 220 | |
| -20° C, Avoid Freeze/Thaw Cycles | |
| Liquid |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction