missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ ALDH1A3 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA515000
This item is not returnable.
View return policy
Description
ALDH1A3 Polyclonal Antibody for Western Blot, IHC (P)
ALDH1A3 are aldehyde dehydrogenase isozymes which are thought to play a major role in the detoxification of aldehydes generated by alcohol metabolism and lipid peroxidation. The enzyme encoded by the ALDH1A3 gene uses retinal as a substrate, either in a free or cellular retinol-binding protein form.
Specifications
| ALDH1A3 | |
| Polyclonal | |
| Unconjugated | |
| ALDH1A3 | |
| acetaldehyde dehydrogenase 6; aldehyde dehydrogenase 1 family member A3; aldehyde dehydrogenase 1 family, member A3; aldehyde dehydrogenase 6; Aldehyde dehydrogenase family 1 member A3; aldehyde dehydrogenase family 1, subfamily A3; aldehyde dehydrogenase-6; ALDH1A3; ALDH1A6; ALDH6; ALDHA1A3; MCOP8; ralDH3; RALDH-3; Retinaldehyde dehydrogenase 3; V1 | |
| Rabbit | |
| Ammonium sulfate precipitation, Size-exclusion - Dialysis | |
| RUO | |
| 220, 56847 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot | |
| 2 mg/mL | |
| PBS with 0.09% sodium azide; pH 7.4 | |
| P47895, Q9JHW9 | |
| ALDH1A3 | |
| KLH conjugated synthetic peptide between 24-51 amino acids from the N-terminal region of human ALDH1A3. | |
| 400 μL | |
| Primary | |
| Human, Mouse | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction