missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ALDH1L1 Antibody (ALDH1L1/7969), DyLight 594, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP3-24234DL594
This item is not returnable.
View return policy
Description
ALDH1L1 Monoclonal antibody specifically detects ALDH1L1 in Human samples. It is validated for Immunohistochemistry (Paraffin)
Specifications
| ALDH1L1 | |
| Monoclonal | |
| DyLight 594 | |
| 10-formyltetrahydrofolate dehydrogenase, aldehyde dehydrogenase 1 family, member L1, aldehyde dehydrogenase family 1 member L1, Cytosolic 10-formyltetrahydrofolate dehydrogenase, DKFZp781N0997, EC 1.5.1.6,10-FTHFDH, FDH, formyltetrahydrofolate dehydrogenase, FTHFD10-fTHF | |
| Recombinant fragment of human ALDH1L1 protein (exact sequence is proprietary) | |
| 0.1 mL | |
| Apoptosis, Cancer, Cell Biology, Cell Cycle and Replication, Endocrinology, Neuroscience, Signal Transduction | |
| 10840 | |
| Store at 4°C in the dark. | |
| IgG2 κ |
| Immunohistochemistry (Paraffin) | |
| ALDH1L1/7969 | |
| 50mM Sodium Borate | |
| Mouse | |
| Protein A or G purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction