missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ALDH1L1 Antibody (OTI3D2), Janelia Fluor™ 549, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-71629JF549
This item is not returnable.
View return policy
Description
ALDH1L1 Monoclonal specifically detects ALDH1L1 in Human, Mouse, Rat, Canine samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence.
Specifications
| ALDH1L1 | |
| Monoclonal | |
| Janelia Fluor 549 | |
| 50mM Sodium Borate with 0.05% Sodium Azide | |
| ALDH1L1 | |
| Full length recombinant protein of human ALDH1L1 (NP_036322) produced in HEK293T cells. | |
| 0.1 mL | |
| 10840.0 | |
| Store at 4°C in the dark. |
| Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence | |
| OTI3D2 | |
| Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence | |
| 10-formyltetrahydrofolate dehydrogenase, aldehyde dehydrogenase 1 family, member L1, aldehyde dehydrogenase family 1 member L1, Cytosolic 10-formyltetrahydrofolate dehydrogenase, DKFZp781N0997, EC 1.5.1.6,10-FTHFDH, FDH, formyltetrahydrofolate dehydrogenase, FTHFD10-fTHF | |
| Mouse | |
| Affinity Purified | |
| Primary | |
| Human, Mouse, Rat, Canine | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction