missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ALDH1L1 Antibody (S06-2E9), Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Novus Biologicals NBP3-19563-100ul
This item is not returnable.
View return policy
Description
ALDH1L1 Monoclonal antibody specifically detects ALDH1L1 in Human, Mouse, Rat samples. It is validated for Western Blot
Specifications
| ALDH1L1 | |
| Monoclonal | |
| 0.3 mg/mL | |
| 50mM Tris-Glycine(pH 7.4), 0.15M NaCl, 40% Glycerol, 0.05% BSA | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Human, Mouse, Rat | |
| Purified |
| Western Blot | |
| S06-2E9 | |
| Unconjugated | |
| 10-formyltetrahydrofolate dehydrogenase, aldehyde dehydrogenase 1 family, member L1, aldehyde dehydrogenase family 1 member L1, Cytosolic 10-formyltetrahydrofolate dehydrogenase, DKFZp781N0997, EC 1.5.1.6,10-FTHFDH, FDH, formyltetrahydrofolate dehydrogenase, FTHFD10-fTHF | |
| A synthetic peptide of human ALDH1L1 (Uniprot # O75891) | |
| 100 μg | |
| Apoptosis, Cancer, Cell Biology, Cell Cycle and Replication, Endocrinology, Neuroscience, Signal Transduction | |
| 10840 | |
| Store at 4°C short term. Store at -20°C long term. Avoid freeze-thaw cycles. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction