Learn More
ALDH1L1 Monoclonal Antibody (OTI3D2), TrueMAB™, OriGene
Mouse Monoclonal Antibody
Brand: Origene Technologies TA501850
This item is not returnable.
View return policy
Description
ALDH1L1 catalyzes the conversion of 10-formyltetrahydrofolate, NADP, and water to tetrahydrofolate, NADPH, and carbon dioxide. The encoded protein belongs to the aldehyde dehydrogenase family and is responsible for formate oxidation in vivo. Deficiencies in this gene can result in an accumulation of formate and subsequent methanol poisoning.Specifications
| ALDH1L1 | |
| Monoclonal | |
| 0.88 mg/mL | |
| PBS with 1% BSA, 50% glycerol and 0.02% sodium azide | |
| O75891 | |
| ALDH1L1, LOC100855730 | |
| Full length human recombit protein of human ALDH1L1 produced in HEK293T cell. | |
| 100 μL | |
| Primary | |
| Canine, Human | |
| Antibody | |
| IgG1 |
| Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot | |
| OTI3D2 | |
| Unconjugated | |
| ALDH1L1 | |
| 10-FTHFDH, 10-fTHF, FDH, FTHFD | |
| Mouse | |
| Affinity Chromatography | |
| RUO | |
| 100855730, 10840 | |
| -20° C, Avoid Freeze/Thaw Cycles | |
| Liquid |