missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ALDH1L1 Monoclonal Antibody (OTI7G6), TrueMAB™, OriGene
Mouse Monoclonal Antibody
Brand: Origene Technologies TA501867
This item is not returnable.
View return policy
Description
ALDH1L1 catalyzes the conversion of 10-formyltetrahydrofolate, NADP, and water to tetrahydrofolate, NADPH, and carbon dioxide. The encoded protein belongs to the aldehyde dehydrogenase family and is responsible for formate oxidation in vivo. Deficiencies in this gene can result in an accumulation of formate and subsequent methanol poisoning.Specifications
| ALDH1L1 | |
| Monoclonal | |
| Unconjugated | |
| ALDH1L1 | |
| 10-FTHFDH, 10-fTHF, FDH, FTHFD | |
| Mouse | |
| Affinity Chromatography | |
| RUO | |
| 10840 | |
| -20° C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| Flow Cytometry, Immunocytochemistry, Immunofluorescence, Western Blot | |
| OTI7G6 | |
| PBS with 1% BSA, 50% glycerol and 0.02% sodium azide | |
| O75891 | |
| ALDH1L1 | |
| Full length human recombit protein of human ALDH1L1 produced in HEK293T cell. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG2a |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction