Learn More
ALDH1L1 Mouse anti-Canine, Human, Clone: OTI6A10, lyophilized, TrueMAB™
Mouse Monoclonal Antibody
Brand: Origene Technologies CF501871
Description
Reconstitute with PBS (pH 7.3) and recommend to perform another round of desalting process using Product No. 7KMWCO
ALDH1L1 catalyzes the conversion of 10-formyltetrahydrofolate, NADP, and water to tetrahydrofolate, NADPH, and carbon dioxide. The encoded protein belongs to the aldehyde dehydrogenase family and is responsible for formate oxidation in vivo. Deficiencies in this gene can result in an accumulation of formate and subsequent methanol poisoning.Specifications
| ALDH1L1 | |
| Monoclonal | |
| Unconjugated | |
| ALDH1L1 | |
| 10-FTHFDH, 10-fTHF, FDH, FTHFD | |
| Mouse | |
| Affinity Chromatography | |
| RUO | |
| 100855730, 10840 | |
| -20° C, Avoid Freeze/Thaw Cycles | |
| Lyophilized |
| Flow Cytometry, Western Blot | |
| OTI6A10 | |
| PBS with 8% trehalose and no preservative; pH 7.3 | |
| O75891 | |
| ALDH1L1, LOC100855730 | |
| Full length human recombit protein of human ALDH1L1 produced in HEK293T cell. | |
| 100 μg | |
| Primary | |
| Canine, Human | |
| Antibody | |
| IgG1 |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.