missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ALDH4A1 Monoclonal Antibody (OTI1H10), TrueMAB™, OriGene
Mouse Monoclonal Antibody
Brand: Origene Technologies TA812719
This item is not returnable.
View return policy
Description
This protein belongs to the aldehyde dehydrogenase family of proteins. This enzyme is a mitochondrial matrix NAD-dependent dehydrogenase which catalyzes the second step of the proline degradation pathway, converting pyrroline-5-carboxylate to glutamate. Deficiency of this enzyme is associated with type II hyperprolinemia, an autosomal recessive disorder characterized by accumulation of delta-1-pyrroline-5-carboxylate (P5C) and proline. Two transcript variants encoding the same protein have been identified for this gene.Specifications
| ALDH4A1 | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with 1% BSA, 50% glycerol and 0.02% sodium azide | |
| P0C2X9, P30038 | |
| ALDH4A1 | |
| Human recombit protein fragment corresponding to amino acids 1-503 of human ALDH4A1 produced in E.coli. | |
| 100 μL | |
| Primary | |
| Canine, Human, Rat | |
| Antibody | |
| IgG1 |
| Immunohistochemistry (Paraffin), Western Blot | |
| OTI1H10 | |
| Unconjugated | |
| ALDH4A1 | |
| ALDH4, P5CD, P5CDh | |
| Mouse | |
| Affinity Chromatography | |
| RUO | |
| 612452, 641316, 8659 | |
| -20° C, Avoid Freeze/Thaw Cycles | |
| Liquid |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction