Learn More
ALDH5A1 Antibody, Allophycocyanin, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand:  Novus Biologicals NBP2-98042APC
Product Code. 18629971
  • € 407.00 / 0.10mL
Description
ALDH5A1 Polyclonal antibody specifically detects ALDH5A1 in Human samples. It is validated for Immunocytochemistry/ Immunofluorescence, Immunohistochemistry (Paraffin)