Learn More
ALDH7A1 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand:  Proteintech 10368-1-AP-150UL
 View more versions of this product
Product Code. 16816713
  • € 410.00 / 150µL
Description
The protein encoded by this gene is a member of subfamily 7 in the aldehyde dehydrogenase gene family. These enzymes are thought to play a major role in the detoxification of aldehydes generated by alcohol metabolism and lipid peroxidation. This particular member has homology to a previously described protein from the green garden pea, the 26g pea turgor protein. It is also involved in lysine catabolism that is known to occur in the mitochondrial matrix. Recent reports show that this protein is found both in the cytosol and the mitochondria, and the two forms likely arise from the use of alternative translation initiation sites. An additional variant encoding a different isoform has also been found for this gene. Mutations in this gene are associated with pyridoxine-dependent epilepsy. Several related pseudogenes have also been identified.
Specifications
Specifications
ALDH7A1
Polyclonal
Unconjugated
ALDH7A1
ALDH7A1, Alpha AASA dehydrogenase, Antiquitin 1, ATQ1, Betaine aldehyde dehydrogenase, EPD, P6c dehydrogenase, PDE
Rabbit
Antigen Affinity Chromatography
RUO
110695, 291450, 501
-20°C
Liquid
Immunohistochemistry (Paraffin), Western Blot
0.15 mg/mL
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3
P49419, Q64057, Q9DBF1
Aldh7a1
ALDH7A1 Fusion Protein Ag0383
150 μL
Primary
Human, Mouse, Rat