missing translation for 'onlineSavingsMsg'
Learn More

Aldicarb Sulfone, TRC

Product Code. 30309619
Click to view available options
Quantity:
100 mg
Unit Size:
100mg
This item is not returnable. View return policy

Product Code. 30309619

Brand: Toronto Research Chemicals TRCA514660100MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Aldicarb-sulfone
CAS 1646-88-4
Molecular Formula C7 H14 N2 O4 S
Quantity 100 mg
Synonym Propionaldehyde, 2-methyl-2-(methylsulfonyl)-, O-(methylcarbamoyl)oxime (7CI,8CI), Carbamic acid, methyl-, O-[[2-methyl-2-(methylsulfonyl)propylidene]amino] deriv. (8CI), 2-Methyl-2-(methylsulfonyl)propionaldehyde O-(methylcarbamoyl)oxime, Aldicarb sulfone, Aldoxycarb, Standak, Sulfocarb, Temik sulfone, UC 21865, UCES, Yangdimiewei
InChI Formula InChI=1S/C7H14N2O4S/c1-7(2,14(4,11)12)5-9-13-6(10)8-3/h5H,1-4H3,(H,8,10)
SMILES CNC(=O)ON=CC(C)(C)S(=O)(=O)C
IUPAC Name [(2-methyl-2-methylsulfonylpropylidene)amino] N-methylcarbamate
Molecular Weight (g/mol) 222.26
Formula Weight 222.0674
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.