missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Aldo-keto Reductase 1C4/AKR1C4 Antibody, Alexa Fluor™ 532, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-97581AF532
This item is not returnable.
View return policy
Description
Aldo-keto Reductase 1C4/AKR1C4 Polyclonal specifically detects Aldo-keto Reductase 1C4/AKR1C4 in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry-Paraffin.
Specifications
| Aldo-keto Reductase 1C4/AKR1C4 | |
| Polyclonal | |
| Immunohistochemistry, Immunohistochemistry-Paraffin | |
| 3-alpha-HSD, AKR1C4, aldo-keto reductase family 1, member C4, C11, CDR, CHDR, DD4, DD-4, HAKRA | |
| Produced in rabbits immunized with E. coli-derived Human Aldo-keto Reductase 1C4/AKR1C4 fragment. | |
| 0.1 mL | |
| Lipid and Metabolism | |
| 1109 | |
| Store at 4°C in the dark. | |
| IgG |
| Immunohistochemistry, Immunohistochemistry (Paraffin) | |
| Alexa Fluor 532 | |
| 50mM Sodium Borate | |
| Rabbit | |
| Antigen Affinity-purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction