Learn More
Invitrogen™ Aldolase A Recombinant Rabbit Monoclonal Antibody (24GB4710)
Rabbit Recombinant Monoclonal Antibody
Brand:  Invitrogen™ MA557330
 View more versions of this product
Product Code. 30274894
  • € 198.00 / 20µL
Description
Aldolase A Recombinant Monoclonal Antibody for Western Blot
Aldolase encodes a member of the class I fructose-bisphosphate aldolase protein family. The encoded protein is a glycolytic enzyme that catalyzes the reversible conversion of fructose-1,6-bisphosphate to glyceraldehyde 3-phosphate and dihydroxyacetone phosphate. Three aldolase isozymes (A, B, and C), encoded by three different genes, are differentially expressed during development. Mutations in this gene have been associated with Glycogen Storage Disease XII, an autosomal recessive disorder associated with hemolytic anemia. Disruption of this gene also plays a role in the progression of multiple types of cancers. Related pseudogenes have been identified on chromosomes 3 and 10.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Aldolase A
Recombinant Monoclonal
Unconjugated
ALDOA
ALDA; Aldo1; Aldo-1; ALDOA; Aldolase 1; aldolase 1, A isoform; aldolase A; aldolase A, fructose-bisphosphate; aldolase, fructose-bisphosphate A; epididymis secretory sperm binding protein Li 87p; fructose-1,6-bisphosphate triosephosphate-lyase; fructose-bisphosphate aldolase A; GSD12; HEL-S-87p; Lung cancer antigen NY-LU-1; muscle-type aldolase; RNALDOG5; unnamed protein product
Rabbit
Affinity chromatography
RUO
11674, 226, 24189
-20°C
Liquid
Western Blot
24GB4710
PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
P04075, P05064, P05065
ALDOA
A synthesized peptide derived from human Aldolase (aa 7-20).
20 μL
Primary
Human, Mouse, Rat