missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Aldolase B Antibody (OTI3C6), CoraFluor™ 1, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-70162CL1
This item is not returnable.
View return policy
Description
Aldolase B Monoclonal antibody specifically detects Aldolase B in Human,Mouse,Rat samples. It is validated for Flow Cytometry,Immunohistochemistry,Western Blot,CyTOF
Specifications
| Aldolase B | |
| Monoclonal | |
| CoraFluor 1 | |
| ALDO2, aldolase B, fructose-bisphosphate, EC 4.1.2.13, fructose-bisphosphatase, fructose-bisphosphate aldolase B | |
| Full length human recombinant protein of human ALDOB(NP_000026) produced in HEK293 cell. | |
| 0.1 mL | |
| Cancer | |
| 229 | |
| Store at 4°C in the dark. Do not freeze. | |
| IgG2b |
| Flow Cytometry, Immunohistochemistry, Western Blot, CyTOF | |
| OTI3C6 | |
| PBS | |
| Mouse | |
| Immunogen affinity purified | |
| RUO | |
| Primary | |
| Human, Mouse, Rat | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction