Learn More
Aldolase C Antibody (E9), Janelia Fluor™ 646, Novus Biologicals™
Mouse Monoclonal Antibody
Brand:  Novus Biologicals NBP2-50056JF646
Product Code. 18756702
  • € 627.00 / 0.10mL
Description
Aldolase C Monoclonal antibody specifically detects Aldolase C in Human, Mouse, Rat, Porcine, Bovine, Chicken, Equine samples. It is validated for Western Blot, Immunofluorescence
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Aldolase C
Monoclonal
Western Blot, Immunocytochemistry/Immunofluorescence
Core ESC Like Genes, Stem Cell Markers
Purified
Western Blot, Immunofluorescence
E9
RUO
Pig