missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Aldolase C Monoclonal Antibody (4A9)
Mouse Monoclonal Antibody
€ 491.00 - € 1704.00
Specifications
| Antigen | Aldolase C |
|---|---|
| Clone | 4A9 |
| Concentration | 1 mg/mL |
| Content And Storage | Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. |
| Applications | Immunohistochemistry, Western Blot, Immunocytochemistry |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
17835307
|
Invitrogen™
MA547327 |
500 μL |
€ 1704.00
500µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
17835317
|
Invitrogen™
MA547426 |
100 μL |
€ 491.00
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Aldolase C Monoclonal Antibody for Western Blot, ICC/IF, IHC
This gene encodes a member of the class I fructose-biphosphate aldolase gene family. Expressed specifically in the hippocampus and Purkinje cells of the brain, the encoded protein is a glycolytic enzyme that catalyzes the reversible aldol cleavage of fructose-1,6-biphosphate and fructose 1-phosphate to dihydroxyacetone phosphate and either glyceraldehyde-3-phosphate or glyceraldehyde, respectively. [provided by RefSeq, Jul 2008].Specifications
| Aldolase C | |
| 1 mg/mL | |
| Immunohistochemistry, Western Blot, Immunocytochemistry | |
| Unconjugated | |
| Mouse | |
| RUO | |
| PBS with 50% glycerol and 5mM sodium azide | |
| P05063, P09117, P09972 | |
| 100512013, 11676, 230, 24191, 504584 | |
| N-terminal sequence MPHSYPALSAEQKKELSDIA (aa 1-20). | |
| Antibody |
| 4A9 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Monoclonal | |
| Liquid | |
| IgG1 | |
| Human, Mouse, Rat, Bovine, Pig | |
| ALDOC | |
| AI847350; ALDC; ALDCAA; Aldo3; ALDOC; Aldolase 3; aldolase C, fructose-biphosphate; aldolase C, fructose-bisphosphate; aldolase, fructose-bisphosphate C; AU040929; Brain-type aldolase; EC 4.1.2.13; F16dip7; fructoaldolase C; fructose-1,6-biphosphate triosephosphate lyase; fructose-bisphosphate aldolase C; RATALDCAA; Scrapie-responsive protein 2; Scrg2; Zebrin II | |
| ALDOC | |
| Primary | |
| Protein G |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title