missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Aldosterone Antibody (BGN/10/42110), Alexa Fluor™ 405, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NB100-62313AF405
This item is not returnable.
View return policy
Description
Aldosterone Monoclonal specifically detects Aldosterone in All Species samples. It is validated for ELISA.
Specifications
| Aldosterone | |
| Monoclonal | |
| Alexa Fluor 405 | |
| 50mM Sodium Borate with 0.05% Sodium Azide | |
| Aldosterone (3-CMO) conjugated to Bovine Serum Albumin. | |
| 0.1 mL | |
| Primary | |
| All species | |
| Purified |
| ELISA | |
| BGN/10/42110 | |
| ELISA | |
| Mouse | |
| Protein A or G purified | |
| RUO | |
| This product is specific for aldosterone and exhibits minimal reactivity with androstenedione, corticosterone, DHEA and desoxycortisol. | |
| Store at 4C in the dark. | |
| IgG1 |
Korrektion af produktindhold
Dit input er vigtigt for os. Udfyld denne formular for at give feedback relateret til indholdet på dette produkt.
Produkttitel
Ser du en mulighed for forbedring?Del en indholdskorrektion