missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Aldosterone Antibody (BGN/10/42110), Alexa Fluor™ 700, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NB100-62313AF700
This item is not returnable.
View return policy
Description
Aldosterone Monoclonal specifically detects Aldosterone in All Species samples. It is validated for ELISA.
Specifications
| Aldosterone | |
| Monoclonal | |
| Alexa Fluor 700 | |
| 50mM Sodium Borate with 0.05% Sodium Azide | |
| Aldosterone (3-CMO) conjugated to Bovine Serum Albumin. | |
| 0.1 mL | |
| Primary | |
| All species | |
| Purified |
| ELISA | |
| BGN/10/42110 | |
| ELISA | |
| Mouse | |
| Protein A or G purified | |
| RUO | |
| This product is specific for aldosterone and exhibits minimal reactivity with androstenedione, corticosterone, DHEA and desoxycortisol. | |
| Store at 4C in the dark. | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction