missing translation for 'onlineSavingsMsg'
Learn More

Aldosterone Antibody (BGN/10/4213), Biotin, Novus Biologicals™

Product Code. 18050804 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18050804 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18050804 Supplier Novus Biologicals Supplier No. NB10062316B

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

Aldosterone Monoclonal specifically detects Aldosterone in All Species samples. It is validated for ELISA.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Aldosterone
Applications ELISA
Classification Monoclonal
Clone BGN/10/4213
Conjugate Biotin
Dilution ELISA
Formulation PBS with 0.05% Sodium Azide
Host Species Mouse
Immunogen Aldosterone conjugated to Bovine Serum Albumin.
Purification Method Protein A purified
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Test Specificity Aldosterone. This antibody shows less than 0.01% cross-reactivity with Androstenedione, Corticosterone, DHEA and Deoxcorticosterone.
Target Species All species
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.