missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Aldosterone Antibody (BGN/10/4213), Janelia Fluor™ 549, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NB100-62316JF549
This item is not returnable.
View return policy
Description
Aldosterone Monoclonal antibody specifically detects Aldosterone in All samples. It is validated for ELISA
Specifications
| Aldosterone | |
| Monoclonal | |
| Janelia Fluor 549 | |
| Mouse | |
| Protein A purified | |
| RUO | |
| Primary | |
| Store at 4°C in the dark. | |
| IgG1 |
| ELISA | |
| BGN/10/4213 | |
| 50mM Sodium Borate | |
| Aldosterone conjugated to Bovine Serum Albumin. | |
| 0.1 mL | |
| Lipid and Metabolism | |
| All species | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction