missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Aldosterone Antibody (BGN/10/42133), DyLight 680, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NB100-62317FR
This item is not returnable.
View return policy
Description
Aldosterone Monoclonal specifically detects Aldosterone in All Species samples. It is validated for ELISA.
Specifications
| Aldosterone | |
| Monoclonal | |
| DyLight 680 | |
| 50mM Sodium Borate with 0.05% Sodium Azide | |
| Aldosterone conjugated to Bovine Serum Albumin. | |
| 0.1 mL | |
| Primary | |
| All species | |
| Purified |
| ELISA | |
| BGN/10/42133 | |
| ELISA | |
| Mouse | |
| Protein G purified | |
| RUO | |
| Aldosterone. NB100-62317 does not cross react with Androstenedione, Corticosterone, DHEA or Desoxycortisol. | |
| Store at 4C in the dark. | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction