missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Aldosterone Antibody, DyLight 550, Novus Biologicals™
Sheep Polyclonal Antibody
Brand: Novus Biologicals NB100-62320R
This item is not returnable.
View return policy
Description
Aldosterone Polyclonal antibody specifically detects Aldosterone in All samples. It is validated for ELISA
Specifications
| Aldosterone | |
| Polyclonal | |
| 50mM Sodium Borate | |
| Made to Broad ALDOSTERONE | |
| 0.1 mL | |
| Lipid and Metabolism | |
| All species | |
| Purified |
| ELISA | |
| DyLight 550 | |
| Sheep | |
| Immunogen affinity purified | |
| RUO | |
| Primary | |
| Store at 4°C in the dark. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction