missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Aldosterone Polyclonal Antibody
GREENER_CHOICE

Product Code. 11549270
Change view
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11549270 100 μL 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 11549270 Supplier Invitrogen™ Supplier No. PA185150

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Sheep Polyclonal Antibody

Sheep anti Aldosterone antibody recognizes aldosterone, a steroid hormone (mineralocorticoid family) produced by the outer-section (zona glomerulosa) of the adrenal cortex in the adrenal gland to regulate the bodies electrolyte and water balance.

Aldosterone is a hormone produced by the adrenal glands that helps regulate blood pressure through increasing both the release of potassium and the uptake of water and sodium by the kidneys. It is essential for sodium conservation in the kidney, salivary glands, sweat glands and colon. It plays a central role in the regulation of the plasma sodium (Na+), the extracellular potassium (K+) and arterial blood pressure. It does so mainly by acting on the mineralocorticoid receptors in the distal tubules and collecting ducts of the nephron. It influences the reabsorption of sodium and excretion of potassium (from and into the tubular fluids, respectively) of the kidney, thereby indirectly influencing water retention or loss, blood pressure and blood volume. When dysregulated, aldosterone is pathogenic and contributes to the development and progression of cardiovascular and renal disease. Aldosterone has exactly the opposite function of the atrial natriuretic hormone secreted by the heart.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Aldosterone
Applications ELISA
Classification Polyclonal
Concentration 5 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.09% sodium azide; pH 7.2
Host Species Sheep
Immunogen Aldosterone conjugated to BSA.
Purification Method Protein G
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Target Species Chemical
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Target Molecule Chemical
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.