Learn More
Alendronate Acid, TRC
Product Code. 30135673
Change view
Click to view available options
Quantity:
1 g
5 g
10 g
Unit Size:
10g
1g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30135673 10 g 10g
30007608 5 g 5g
30004975 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 30135673 Supplier Toronto Research Chemicals Supplier No. TRCA52126310G
 Restricted Item
This item requires a price update prior to being ordered.
Request a Price for this item here.
missing translation for 'validMainframeMsg'
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 66376-36-1
Molecular Formula C4H13NO7P2
Molecular Weight (g/mol) 249.1
Synonym P,P'-(4-Amino-1-hydroxybutylidene)bis[phosphonic Acid], 4-Amino-1-hydroxybutane-1,1-bisphosphonic Acid, 4-Amino-1-hydroxybutane-1,1-diphosphonate, 4-Amino-1-hydroxybutane-1,1-diphosphonic Acid, 4-Amino-1-hydroxybutane-1,1-diyldiphosphonic Acid, 4-Amino-1-hydroxybutylidene-1,1-bis(phosphonic Acid), ABDP, Alendronic Acid(4-amino-1-hydroxybutylidene)bis-, BPH 1, Marvil, (4-Amino-1-hydroxybutylidene)bis-phosphonic Acid
IUPAC Name (4-amino-1-hydroxy-1-phosphonobutyl)phosphonic acid
SMILES C(CC(O)(P(=O)(O)O)P(=O)(O)O)CN
Specifications
Quantity 10 g
InChI Formula InChI=1S/C4H13NO7P2/c5-3-1-2-4(6,13(7,8)9)14(10,11)12/h6H,1-3,5H2,(H2,7,8,9)(H2,10,11,12)
Formula Weight 249.0167
Chemical Name or Material Alendronate Acid