missing translation for 'onlineSavingsMsg'
Learn More

Molecular Probes™ Alexa Fluor™ 488 5-SDP Ester (Alexa Fluor™ 488 Sulfodichlorophenol Ester)

Product Code. 10256683
Click to view available options
Quantity:
1 mg
Unit Size:
1mg
This item is not returnable. View return policy

Product Code. 10256683

Brand: Molecular Probes™ A30052

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Alexa Fluor™ 488 5-SDP ester (Alexa Fluor™ 488 sulfodichlorophenol ester)

The SDP (sulfodicholorphenol) ester is currently the most hydrolytically stable amine-reactive moiety available from Invitrogen. Conjugates produced with the Alexa Fluor™ 488 5-SDP ester produce the same strong amide bond between the dye and the compound of interest as succinimidyl (SE or NHS) and tetrafluorophenyl (TFP) esters, but with the improved stability in water and buffers, the SDP ester can potentially offer increased control and consistency in these reactions compared to the NHS and TFP esters.
TRUSTED_SUSTAINABILITY

Specifications

Quantity 1 mg
Product Type Sulfodichlorophenol Ester
Content And Storage Store in freezer (-5 to -30°C) and protect from light.
Chemical Reactivity Amine
Shipping Condition Room Temperature
Reactive Moiety Active Ester, SDP Ester
Label or Dye Alexa Fluor™ 488
Label Type Alexa Fluor
Product Line Alexa Fluor

For Research Use Only. Not for use in diagnostic procedures.

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.