Learn More
Invitrogen™ Alexa Fluor™ 488 Oligonucleotide Amine Labeling Kit
Product Code. 10788604
Click to view available options
Quantity:
1 kit
Unit Size:
Each
Product Code. 10788604
Brand: Invitrogen™ A20191
Alexa Fluor™ 488 Oligonucleotide Amine Labeling Kit
The Alexa Fluor 488 Oligonucleotide Amine Labeling Kit provides a simple method for labeling amine-modified oligonucleotides with our bright, photostable, green-fluorescent Alexa Fluor 488 dye (which is spectrally similar to fluorescein). Oligonucleotides labeled with this kit can be used as primers and as probes in hybridization experiments but should not be used for automated DNA sequencing since the Alexa Fluor dye in the kit is supplied as mixed isomers. The kits have been optimized for labeling reactions with 50μg of a 5'-amine—modified oligonucleotide, 18 to 24 bases in length.
TRUSTED_SUSTAINABILITY
Specifications
Content And Storage Store in freezer (-5 to -30°C) and protect from light.
Detection Method Fluorescence
Format Kit
Label or Dye Alexa Fluor 488
Final Product Type Oligos (Labeled)
Labeling Target Oligos
Includes Label or Dye Yes
Labeling Method Direct Labeling
Product Line Alexa Fluor
Product Type Olionucleotide Amine Labeling Kit
Quantity 1 kit
Shipping Condition Room Temperature