Learn More
Alfacalcidol, Mikromol™
Click to view available options
Quantity:
50 mg
Unit Size:
50mg
Description
Alfacalcidol
Specifications
Specifications
| Chemical Name or Material | Alfacalcidol |
| CAS | 41294-56-8 |
| Analyte or Component Names | Alfacalcidol |
| Shipping Condition | Ice pack (-20°C) |
| Molecular Formula | C27 H44 O2 |
| Certifications/Compliance | ISO 9001 |
| Synonym | Alfacalcidol, 1,3-Cyclohexanediol, 5-[(2E)-2-[(1R,3aS,7aR)-1-[(1R)-1,5-dimethylhexyl]octahydro-7a-methyl-4H-inden-4-ylidene]ethylidene]-4-methylene-, (1R,3R)-, (1R,3R)-5-[(2E)-2-[(1R,3aS,7aR)-1-[(1R)-1,5-Dimethylhexyl]octahydro-7a-methyl-4H-inden-4-ylidene]ethylidene]-4-methylene-1,3-cyclohexanediol, (1S)-Hydroxycalciol, 1-Hydroxycholecalciferol, 1-Hydroxyvitamin D3, Alfarol, Alpha D3, Alphacalcidol, Alpharol, Bondiol, EinsAlpha, Etalpha, Oxydevit, Tevabone, Un Alfa, Un Alpha, (5Z,7E)-9,10-Secocholesta-5,7,10(19)-triene-1alpha,3beta-diol, 9,10-Secocholesta-5,7,10(19)-triene-1,3-diol, (1alpha,3beta,5Z,7E)- (9CI), 1alpha(OH)D3, 1alpha-Hydroxycholecalciferol, 1alpha-Hydroxyvitamin D3, alpha-Calcidol |
| InChI Formula | InChI=1S/C27H44O2/c1-18(2)8-6-9-19(3)24-13-14-25-21(10-7-15-27(24,25)5)11-12-22-16-23(28)17-26(29)20(22)4/h11-12,18-19,23-26,28-29H,4,6-10,13-17H2,1-3,5H3/b21-11+,22-12-/t19-,23-,24-,25+,26+,27-/m1/s1 |
| SMILES | CC(C)CCC[C@@H](C)[C@H]1CC[C@H]2\C(=C\C=C/3\C[C@@H](O)C[C@H](O)C3=C)\CCC[C@]12C |
| IUPAC Name | (1R,3S,5Z)-5-[(2E)-2-[(1R,3aS,7aR)-7a-methyl-1-[(2R)-6-methylheptan-2-yl]-2,3,3a,5,6,7-hexahydro-1H-inden-4-ylidene]ethylidene]-4-methylidenecyclohexane-1,3-diol |
| Show More |