missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ ALG6 Polyclonal Antibody
GREENER_CHOICE

Product Code. 17814520
Change view
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
17814520 100 μL 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 17814520 Supplier Invitrogen™ Supplier No. PA5140000

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

Gently mix before use. Optimal concentrations and conditions for each application should be determined by the user. Immunogen sequence: SGAGKPPMFG DYEAQRHWQE ITFNLPVKQW YFNSSDNNLQ YWGLDYPPLT A Highest antigen sequence indentity to the following orthologs: Mouse - 96%, Rat - 98%.

ALG6 is a member of the ALG6/ALG8 glucosyltransferase family. It catalyzes the addition of the first glucose residue to the growing lipid-linked oligosaccharide precursor of N-linked glycosylation. Mutations in this gene encoding ALG6 are associated with congenital disorders of glycosylation type Ic.This gene encodes a member of the ALG6/ALG8 glucosyltransferase family. The encoded protein catalyzes the addition of the first glucose residue to the growing lipid-linked oligosaccharide precursor of N-linked glycosylation. Mutations in this gene are associated with congenital disorders of glycosylation type Ic.
TRUSTED_SUSTAINABILITY

Specifications

Antigen ALG6
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 0.05 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene ALG6
Gene Accession No. Q9Y672
Gene Alias Alg6; ALG6 alpha-1,3-glucosyltransferase; ALG6, alpha-1,3-glucosyltransferase; asparagine-linked glycosylation 6 (alpha-1,3,-glucosyltransferase); asparagine-linked glycosylation 6 homolog (S. cerevisiae, alpha-1,3-glucosyltransferase); asparagine-linked glycosylation 6 homolog (yeast, alpha-1,3,-glucosyltransferase); asparagine-linked glycosylation 6 homolog (yeast, alpha-1,3-glucosyltransferase); asparagine-linked glycosylation 6, alpha-1,3-glucosyltransferase homolog; asparagine-linked glycosylation protein 6 homolog; CDG1C; dolichyl pyrophosphate Man9GlcNAc2 alpha-1,3-glucosyltransferase; Dolichyl pyrophosphate Man9GlcNAc2 alpha-1,3-glucosyltransferase-like protein; dolichyl-P-Glc:Man(9)GlcNAc(2)-PP-dolichol alpha- 1->3-glucosyltransferase; dolichyl-P-Glc:Man9GlcNAc2-PP-dolichyl glucosyltransferase; dolichyl-P-Glc:Man9GlcNAc2-PP-dolichylglucosyltransferase; Dol-P-Glc:Man(9)GlcNAc(2)-PP-Dol alpha-1,3-glucosyltransferase; E230028F23Rik; Man(9)GlcNAc(2)-PP-Dol alpha-1,3-glucosyltransferase; My046; zgc:100813
Gene Symbols ALG6
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human ALG6. Recombinant protein control fragment (Product #RP-109535).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 29929
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.