missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ ALG8 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA550317
This item is not returnable.
View return policy
Description
The antibody detects endogenous levels of total ALG8 protein.
Adds the second glucose residue to the lipid-linked oligosaccharide precursor for N-linked glycosylation. Transfers glucose from dolichyl phosphate glucose (Dol-P-Glc) onto the lipid-linked oligosaccharide Glc(1)Man(9)GlcNAc(2)-PP-Dol.
Specifications
| ALG8 | |
| Polyclonal | |
| Unconjugated | |
| ALG8 | |
| AI447372; alg8; ALG8 alpha-1,3-glucosyltransferase; ALG8, alpha-1,3-glucosyltransferase; asparagine-linked glycosylation 8 (alpha-1,3-glucosyltransferase); asparagine-linked glycosylation 8 alpha-13-glucosyltransferase-like protein; asparagine-linked glycosylation 8 homolog; asparagine-linked glycosylation 8 homolog (S. cerevisiae, alpha-1,3-glucosyltransferase); asparagine-linked glycosylation 8 homolog (yeast, alpha-1,3-glucosyltransferase); asparagine-linked glycosylation 8, alpha-1,3-glucosyltransferase homolog; asparagine-linked glycosylation protein 8 homolog; CDG1H; dolichyl pyrophosphate Glc1Man9GlcNAc2 alpha-1,3-glucosyltransferase; dolichyl-P-Glc:Glc(1)Man(9)GlcNAc(2)-PP-dolichol alpha- 1->3-glucosyltransferase; dolichyl-P-Glc:Glc1Man9GlcNAc2-PP-dolichyl glucosyltransferase; dolichyl-P-glucose:Glc1Man9GlcNAc2-PP-dolichyl-alpha-1,3-glucosyltransferase; Dol-P-Glc:Glc(1)Man(9)GlcNAc(2)-PP-dolichyl alpha-1,3-glucosyltransferase; Gm1089; HUSSY-02; hypothetical protein LOC327601; probable dolichyl pyrophosphate Glc1Man9GlcNAc2 alpha-1,3-glucosyltransferase; putative dolichyl pyrophosphate Glc1Man9GlcNAc2 alpha-1,3-glucosyltransferase-like protein; si:rp71-1p14.2; Unknown (protein for MGC:142391); wu:fi19e09; wu:fi19f07; zgc:112507 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 79053 | |
| -20°C | |
| Liquid |
| Immunohistochemistry (Paraffin) | |
| 1.2 mg/mL | |
| PBS with 40% glycerol and 0.05% sodium azide; pH 7.4 | |
| Q9BVK2 | |
| ALG8 | |
| Fusion protein corresponding to a region derived from internal residues of human ALG8, alpha-1,3-glucosyltransferase. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction