missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Alginate Monoclonal Antibody (3G4-1F5), APC

Mouse Monoclonal Antibody
Brand: Invitrogen™ MA545315
This item is not returnable.
View return policy
Description
1 μg/mL of MA5-45315 was sufficient for detection of 150 ng of alginate-conjugated BSA by colorimetric immunoblot analysis using Goat anti-mouse IgG:HRP as the secondary antibody. Binds selectively to a BSA-conjugated alginate, but not to unconjugated BSA.
Sodium alginate is used in biological experiments for the immobilization of cells and encapsulation due to its biocompatibility and simple gelation with divalent cations such as Ca2+. Studies suggest that preparation of alginate microspheres will sustain protein delivery within tissue scaffolds, as well as in many other cell types.
Specifications
| Alginate | |
| Monoclonal | |
| 1 mg/mL | |
| 2.48mM MES, 95.64mM phosphate with 0.5M EDTA and no preservative; pH 7.4 | |
| Mouse | |
| Protein G | |
| RUO | |
| Chemical | |
| Antibody | |
| IgG1 κ |
| Immunohistochemistry, Western Blot | |
| 3G4-1F5 | |
| APC | |
| Algin; Alginic Acid; Sodium Alginate | |
| Sodium Alginate conjugated to KLH. | |
| 100 μg | |
| Primary | |
| 4°C | |
| Liquid | |
| Chemical |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction