Learn More
Invitrogen™ Alirocumab Recombinant Monoclonal Antibody
Recombinant Monoclonal Antibody
Brand: Invitrogen™ MA558741
This item is not returnable.
View return policy
Description
For reconstitution, add sterile, distilled water to achieve a final antibody concentration of 1 mg/mL. Gently shake to solubilize the protein completely. Do not vortex. Reconstituted products should be stored at -80 °.
Alirocumab is a biopharmaceutical that obtained FDA approval in July 2015 as a second line treatment for high cholesterol in adults whose LDL-cholesterol (LDL-C) is not controlled by the combination of diet and statin treatment. It is a human monoclonal antibody part of the family of the PCSK9 inhibitors which are a novel class of anticholesterol therapeutics. From this family, it was the first agent to receive FDA approval. The FDA approval was contingent on the completion of further clinical trials to better determine efficacy and safety. PCSK9 inhibition facilitates more LDL-C clearance from the blood.
Specifications
| Alirocumab | |
| Recombinant Monoclonal | |
| Unconjugated | |
| REGN727; SAR-236553 | |
| Protein A | |
| RUO | |
| Human | |
| Antibody | |
| IgG1 |
| ELISA, Flow Cytometry, Functional Assay, Surface Plasmon Resonance | |
| 1 mg/mL | |
| 25mM histidine with 8% sucrose, 0.01% Tween 80 and no preservative; pH 6.2 | |
| Human | |
| 100 μg | |
| Primary | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Lyophilized |