missing translation for 'onlineSavingsMsg'
Learn More

Alisertib, MedChemExpress

Product Code. p-200099805
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mg
10 mg
50 mg
100 mg
200 mg
Unit Size:
100mg
10mg
200mg
50mg
5mg
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16497037 5 mg 5mg
16495236 100 mg 100mg
16471006 200 mg 200mg
16470647 50 mg 50mg
16414328 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
This item is not returnable. View return policy
Product Code. 16497037 Supplier Medchem Express Supplier No. HY10971

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress Alisertib (MLN 8237) is an orally active and selective Aurora A kinase inhibitor (IC50=1.2 nM), which binds to Aurora A kinase resulting in mitotic spindle abnormalities, mitotic accumulation. Alisertib (MLN 8237) induces apoptosis and autophagy through targeting the AKT/mTOR/AMPK/p38 pathway in leukemic cells. Antitumor activity.

Alisertib (MLN 8237) is an orally active and selective Aurora A kinase inhibitor (IC50=1.2 nM), which binds to Aurora A kinase resulting in mitotic spindle abnormalities, mitotic accumulation. Alisertib (MLN 8237) induces apoptosis and autophagy through targeting the AKT/mTOR/AMPK/p38 pathway in leukemic cells. Antitumor activity.

Specifications

Chemical Name or Material Alisertib
CAS 1028486-01-2
Color Light Nude
Purity Grade Notes Research
Molecular Formula C27H20ClFN4O4
Quantity 5 mg
Synonym MLN 8237
Solubility Information DMSO : 25 mg/mL (48.18 mM; Need ultrasonic)
SMILES O=C(C1=CC=C(C=C1OC)NC2=NC=C3CN=C(C4=CC(Cl)=CC=C4C3=N2)C5=C(C=CC=C5F)OC)O
Molecular Weight (g/mol) 518.92
Formula Weight 518.92
Percent Purity 95.0%
Grade Research
Physical Form Solid
For Use With (Application) Cancer-Kinase/protease
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.