Learn More
Alismoxide, MedChemExpress
Description
Alismoxide is a natural product.
Specifications
Specifications
| Chemical Name or Material | Alismoxide |
| CAS | 87701-68-6 |
| Color | White |
| Purity Grade Notes | Research |
| Molecular Formula | C15H26O2 |
| Quantity | 10 mg |
| Synonym | (+)-Alismoxide |
| Solubility Information | DMSO : ≥ 31 mg/mL (130.05 mM) |
| SMILES | O[C@@]1(C)CC[C@@]2([H])[C@@](O)(C)CCC(C(C)C)=C[C@]12[H] |
| Molecular Weight (g/mol) | 238.37 |
| Show More |
Research purposes only