missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Alisol B 23-Acetate, TRC
Click to view available options
Quantity:
10 mg
Unit Size:
10mg
Specifications
Specifications
| Chemical Name or Material | Alisol B Acetate |
| CAS | 26575-95-1 |
| Molecular Formula | C32 H50 O5 |
| Quantity | 10 mg |
| Synonym | 23-O-Acetylalisol B, Alisol B 23-acetate |
| InChI Formula | InChI=1S/C32H50O5/c1-18(16-23(36-19(2)33)27-29(5,6)37-27)20-10-14-31(8)21(20)17-22(34)26-30(7)13-12-25(35)28(3,4)24(30)11-15-32(26,31)9/h18,22-24,26-27,34H,10-17H2,1-9H3/t18-,22+,23+,24+,26+,27-,30+,31+,32+/m1/s1 |
| SMILES | C[C@H](C[C@H](OC(=O)C)[C@H]1OC1(C)C)C2=C3C[C@H](O)[C@H]4[C@@]5(C)CCC(=O)C(C)(C)[C@@H]5CC[C@]4(C)[C@@]3(C)CC2 |
| IUPAC Name | [(1S,3R)-1-[(2R)-3,3-dimethyloxiran-2-yl]-3-[(5R,8S,9S,10S,11S,14R)-11-hydroxy-4,4,8,10,14-pentamethyl-3-oxo-1,2,5,6,7,9,11,12,15,16-decahydrocyclopenta[a]phenanthren-17-yl]butyl] acetate |
| Molecular Weight (g/mol) | 514.74 |
| Formula Weight | 514.3658 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction