missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Alizapride Hydrochloride, TRC
Click to view available options
Quantity:
50 mg
Unit Size:
50mg
Specifications
Specifications
| Chemical Name or Material | Alizapride Hydrochloride |
| CAS | 59338-87-3 |
| Molecular Formula | C16 H21 N5 O2 . Cl H |
| Quantity | 50 mg |
| Synonym | 1H-Benzotriazole-5-carboxamide, 6-methoxy-N-[[1-(2-propen-1-yl)-2-pyrrolidinyl]methyl]-, hydrochloride (1:1), 1H-Benzotriazole-5-carboxamide, 6-methoxy-N-[[1-(2-propenyl)-2-pyrrolidinyl]methyl]-, monohydrochloride (9CI), Alizapride hydrochloride, Limican, Nausilen, Plitican, Vergentan |
| InChI Formula | InChI=1S/C16H21N5O2.ClH/c1-3-6-21-7-4-5-11(21)10-17-16(22)12-8-13-14(19-20-18-13)9-15(12)23-2;/h3,8-9,11H,1,4-7,10H2,2H3,(H,17,22)(H,18,19,20);1H |
| SMILES | Cl.COc1cc2[nH]nnc2cc1C(=O)NCC3CCCN3CC=C |
| IUPAC Name | 6-methoxy-N-[(1-prop-2-enylpyrrolidin-2-yl)methyl]-1H-benzotriazole-5-carboxamide;hydrochloride |
| Molecular Weight (g/mol) | 351.83 |
| Formula Weight | 351.1462 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction