missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ALK-1 Antibody (024), mFluor Violet 500 SE, Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Novus Biologicals NBP2-89255MFV500
This item is not returnable.
View return policy
Description
ALK-1 Monoclonal antibody specifically detects ALK-1 in Human samples. It is validated for ELISA, ELISA
Specifications
| ALK-1 | |
| Monoclonal | |
| mFluor Violet 500 SE | |
| 50mM Sodium Borate | |
| Rabbit | |
| Protein A purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
| ELISA, Sandwich ELISA | |
| 024 | |
| ELISA, Sandwich ELISA Detection | |
| activin A receptor type II-like 1, activin A receptor, type II-like kinase 1, Activin receptor-like kinase 1, ACVRLK1ORW2, ALK-1, ALK1TGF-B superfamily receptor type I, EC 2.7.11, HHT, HHT2EC 2.7.11.30, serine/threonine-protein kinase receptor R3, SKR3, TSR-I | |
| This antibody was obtained from a rabbit immunized with purified, recombinant Human ALK-1 (Accession#: NP_000011.2; Met1-Gln118). | |
| 0.1 mL | |
| Protein Kinase | |
| 94 | |
| Store at 4C in the dark. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction