Learn More
ALK-1 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand:  Novus Biologicals NBP1-30982
Product Code. 18716893
  • ONLINE EXCLUSIVE PRICE
    € 618.00
    € 583.80 / 100µL
    Save € 34.20
    5% Off
Description
ALK-1 Polyclonal specifically detects ALK-1 in Human, Mouse samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
activin A receptor type II-like 1, activin A receptor, type II-like kinase 1, Activin receptor-like kinase 1, ACVRLK1ORW2, ALK-1, ALK1TGF-B superfamily receptor type I, EC 2.7.11, HHT, HHT2EC 2.7.11.30, serine/threonine-protein kinase receptor R3, SKR3, TSR-I
Rabbit
56 kDa
Human, Mouse
ACVRL1
Carrier-protein conjugated synthetic peptide encompassing a sequence within the C-terminus region of human ACVRL1. The exact sequence is proprietary.