missing translation for 'onlineSavingsMsg'
Learn More

ALK/CD246 Antibody (ALK/1032), Biotin, Novus Biologicals™

Mouse Monoclonal Antibody

Brand:  Novus Biologicals NBP3-08772B

Product Code. 18630909

  • € 643.00 / 100µL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

ALK/CD246 Monoclonal antibody specifically detects ALK/CD246 in Human samples. It is validated for Western Blot
TRUSTED_SUSTAINABILITY
Specifications

Specifications

ALK/CD246
Monoclonal
Biotin
ALK tyrosine kinase receptor, Anaplastic lymphoma kinase, anaplastic lymphoma kinase (Ki-1), anaplastic lymphoma receptor tyrosine kinase, CD246, CD246 antigen, EC 2.7.10.1, NBLST3
Recombinant fragment (around aa 220-335) of human ALK/CD246 protein (exact sequence is proprietary) (Uniprot: Q9UM73)
100 μg
Protein Kinase, Signal Transduction
238
Store at 4°C in the dark.
IgG2b κ
Western Blot
ALK/1032
PBS
Mouse
Protein A or G purified
RUO
Primary
Human
Purified
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.