Learn More
Inhibin alpha Antibody (INHA/4266), PerCP, Novus Biologicals™
Mouse Monoclonal Antibody
Brand:  Novus Biologicals NBP3-08780PCP
Product Code. 18674308
  • € 702.00 / 0.10mL
Description
Inhibin alpha Monoclonal antibody specifically detects Inhibin alpha in Human samples. It is validated for Immunohistochemistry (Paraffin)
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Inhibin alpha
Monoclonal
PerCP
A-inhibin subunit, INHA, inhibin alpha chain, inhibin, alpha
A synthetic peptide (around aa73-96) of alpha-subunit of human Inhibin alpha protein (exact sequence is proprietary) (Uniprot: P05111 )
0.1 mL
Apoptosis, Cancer
3623
Store at 4°C in the dark.
IgG2a
Immunohistochemistry (Paraffin)
INHA/4266
PBS
Mouse
Protein A or G purified