missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ALK/CD246 Mouse anti-Human, Clone: ALK/1504, Novus Biologicals™
Mouse Monoclonal Antibody
€ 508.00
Specifications
| Antigen | ALK/CD246 |
|---|---|
| Clone | ALK/1504 |
| Concentration | 0.2 mg/mL |
| Dilution | Immunohistochemistry-Paraffin 1 to 2 μg/mL |
| Applications | Flow Cytometry, Immunohistochemistry, Immunohistochemistry (Paraffin), Immunocytochemistry/Immunofluorescence |
Description
ALK/CD246 Monoclonal specifically detects ALK/CD246 in Human samples. It is validated for Immunohistochemistry-Paraffin.Specifications
| ALK/CD246 | |
| 0.2 mg/mL | |
| Flow Cytometry, Immunohistochemistry, Immunohistochemistry (Paraffin), Immunocytochemistry/Immunofluorescence | |
| Unconjugated | |
| Mouse | |
| Phospho Specific, Protein Kinase, Signal Transduction | |
| 10 mM PBS with 0.05% BSA | |
| 238 | |
| IgG2b κ | |
| Protein A or G purified |
| ALK/1504 | |
| Immunohistochemistry-Paraffin 1 to 2 μg/mL | |
| Monoclonal | |
| Purified | |
| RUO | |
| Human | |
| ALK tyrosine kinase receptor, Anaplastic lymphoma kinase, anaplastic lymphoma kinase (Ki-1), anaplastic lymphoma receptor tyrosine kinase, CD246, CD246 antigen, EC 2.7.10.1, NBLST3 | |
| Recombinant human ALK/CD246 protein fragment (aa200-335) | |
| Primary | |
| Store at 4°C. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title