missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ALK Monoclonal Antibody (OTI10D7), TrueMAB™, OriGene
Mouse Monoclonal Antibody
Brand: Origene Technologies TA801306S
This item is not returnable.
View return policy
Description
ALK Monoclonal antibody specifically detects ALK in Human samples. It is validated for Western BlotSpecifications
| ALK | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with 1% BSA, 50% glycerol and 0.02% sodium azide | |
| Q9UM73 | |
| Mouse | |
| Affinity chromatography | |
| RUO | |
| 238 | |
| -20° C, Avoid Freeze/Thaw Cycles | |
| IgG2b |
| Western Blot | |
| OTI10D7 | |
| Unconjugated | |
| ALK | |
| Alk; ALK tyrosine kinase receptor; ALK/NPM1 fusion gene; anaplastic lymphoma kinase; Anaplastic lymphoma kinase Ki1; Anaplastic Lymphoma Kinase p80; anaplastic lymphoma receptor tyrosine kinase; CD246; CD246 antigen; EC 2.7.10.1; kinase ALK; mutant anaplastic lymphoma kinase; NBLST3; npm-alk; p80; Tcrz; TFG/ALK | |
| Human recombinant protein fragment corresponding to amino acids 1300-1620 of human ALK produced in E.coli. | |
| 30 μL | |
| Primary | |
| Human | |
| Liquid |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction