missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ALK Monoclonal Antibody (OTI40F10), TrueMAB™, OriGene
Mouse Monoclonal Antibody
Brand: Origene Technologies TA801407
This item is not returnable.
View return policy
Description
The 2;5 chromosomal translocation is frequently associated with anaplastic large cell lymphomas. The translocation creates a fusion gene consisting of the ALK gene and the nucleophosmin gene: the 3′ half of ALK, derived from chromosome 2, is fused to the 5′ portion of NPM from chromosome 5. A recent study shows that the product of the NPM-ALK fusion gene is oncogenic. The deduced amino acid sequences reveal that ALK is a novel receptor protein-tyrosine kinase having a putative transmembrane domain and an extracellular domain. These sequences are absent in the product of the transforming NPM-ALK gene. ALK shows the greatest sequence similarity to LTK. ALK plays an important role in the development of the brain and exerts its effects on specific neurons in the nervous system.Specifications
| ALK | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with 1% BSA, 50% glycerol and 0.02% sodium azide | |
| Q9UM73 | |
| ALK | |
| Human recombit protein fragment corresponding to amino acids 1300-1620 of human ALK produced in E.coli. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG1 |
| Western Blot | |
| OTI40F10 | |
| Unconjugated | |
| ALK | |
| CD246, NBLST3 | |
| Mouse | |
| Affinity Chromatography | |
| RUO | |
| 238 | |
| -20° C, Avoid Freeze/Thaw Cycles | |
| Liquid |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction