Learn More
ALK Monoclonal Antibody (OTI5H2), TrueMAB™, OriGene
Mouse Monoclonal Antibody
Brand:  Origene Technologies TA800740S
Encompass_Preferred
 View more versions of this product
Product Code. 17598882
  • POA / 30µL
This item requires a price update prior to being ordered.
Request a Price for this item here.
View alternative products »
Description
Description
ALK Monoclonal antibody specifically detects ALK in Human samples. It is validated for Western Blot
Specifications
Specifications
ALK
Monoclonal
1 mg/mL
PBS with 1% BSA, 50% glycerol and 0.02% sodium azide
Q9UM73
Mouse
Affinity chromatography
RUO
238
-20° C, Avoid Freeze/Thaw Cycles
IgG1
Western Blot
OTI5H2
Unconjugated
ALK
Alk; ALK tyrosine kinase receptor; ALK/NPM1 fusion gene; anaplastic lymphoma kinase; Anaplastic lymphoma kinase Ki1; Anaplastic Lymphoma Kinase p80; anaplastic lymphoma receptor tyrosine kinase; CD246; CD246 antigen; EC 2.7.10.1; kinase ALK; mutant anaplastic lymphoma kinase; NBLST3; npm-alk; p80; Tcrz; TFG/ALK
Human recombinant protein fragment corresponding to amino acids 1060-1620 of human ALK produced in SF9 cell.
30 μL