missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Alkaline Phosphatase/ALPP Antibody (ALPP/238), Alexa Fluor™ 647, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-47990AF647
This item is not returnable.
View return policy
Description
Alkaline Phosphatase/ALPP Monoclonal antibody specifically detects Alkaline Phosphatase/ALPP in Human samples. It is validated for ELISA
Specifications
| Alkaline Phosphatase/ALPP | |
| Monoclonal | |
| Alexa Fluor 647 | |
| Alkaline phosphatase Regan isozyme, alkaline phosphatase, placental, alkaline phosphatase, placental (Regan isozyme), alkaline phosphatase, placental type, alkaline phosphomonoesterase, ALP, EC 3.1.3.1, FLJ61142, glycerophosphatase, PALP, Placental alkaline phosphatase 1, PLAP, PLAP-1, Regan isozyme | |
| Recombinant full-length human Alkaline Phosphatase/ALPP protein (Uniprot: P05187) | |
| 0.1 mL | |
| Cancer, Ovarian Carcinoma Cell Markers, Stem Cell Markers | |
| 250 | |
| Store at 4°C in the dark. | |
| IgG2a κ |
| ELISA | |
| ALPP/238 | |
| 50 mM Sodium Borate | |
| Mouse | |
| Protein A or G purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction