missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Alkaline Phosphatase/ALPP Antibody (ALPP/8112R), Novus Biologicals™
Rabbit Monoclonal Antibody
€ 268.00 - € 496.00
Specifications
| Antigen | Alkaline Phosphatase/ALPP |
|---|---|
| Clone | ALPP/8112R |
| Concentration | 0.2 mg/mL |
| Dilution | Immunohistochemistry-Paraffin : 1-2 μg/mL |
| Applications | Immunohistochemistry (Paraffin) |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18621565
|
Novus Biologicals
NBP3-23813-20ug |
20 μg |
€ 268.00
20µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18690535
|
Novus Biologicals
NBP3-23813-100ug |
100 μg |
€ 496.00
100µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Alkaline Phosphatase/ALPP Monoclonal antibody specifically detects Alkaline Phosphatase/ALPP in Human samples. It is validated for Immunohistochemistry (Paraffin)Specifications
| Alkaline Phosphatase/ALPP | |
| 0.2 mg/mL | |
| Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| Rabbit | |
| Cancer, Ovarian Carcinoma Cell Markers, Phospho Specific, Stem Cell Markers | |
| 10 mM PBS with 0.05% BSA | |
| 250 | |
| IgG κ | |
| Protein A or G purified |
| ALPP/8112R | |
| Immunohistochemistry-Paraffin : 1-2 μg/mL | |
| Monoclonal | |
| Purified | |
| RUO | |
| Human | |
| Alkaline phosphatase Regan isozyme, alkaline phosphatase, placental, alkaline phosphatase, placental (Regan isozyme), alkaline phosphatase, placental type, alkaline phosphomonoesterase, ALP, EC 3.1.3.1, FLJ61142, glycerophosphatase, PALP, Placental alkaline phosphatase 1, PLAP, PLAP-1, Regan isozyme | |
| Recombinant full-length human Alkaline Phosphatase/ALPP protein | |
| Primary | |
| Store at 4°C. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title